Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Rabbit anti-Human, Clone: ITGB3/2166R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30755820UG
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Integrin beta 3/CD61 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Recombinant human Integrin beta 3/CD61 protein fragment (aa 385-490) (exact sequence is proprietary) | |
20 μg | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers | |
3690 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
ITGB3/2166R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title