Learn More
Integrin beta-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 183091AP
Catalog No. 50-173-2106
Add to cart
Description
Description
ITGB3 is involved in Glanzmann thrombasthenia (GT), a common inherited disease of platelet aggregation. It is characterized by mucocutaneous bleeding of mild-to-moderate severity. GT has been classified clinically into types I and II. In type I, platelets show absence of the glycoprotein IIb-IIIa complexes at their surface and lack fibrinogen and clot retraction capability. In type II, the platelets express the GPIIb-IIIa complex at reduced levels, have detectable amounts of fibrinogen, and have low or moderate clot retraction capability. The disease is caused by mutations affecting the gene represented in this entry.
Specifications
Specifications
Integrin beta-3
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54890, P05106
CD61, GP3A, GPIIIa, Integrin beta 3, Integrin beta-3, ITGB3
Rabbit
IgG
150 μL
-20°C
Primary
16416, 29302, 3690
Immunoprecipitation, Western Blot
Unconjugated
ITGB3
Liquid
ITGB3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat