Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB5 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550991

Catalog No. PA550991


Add to cart

Description

Description

Integrin beta 5 Polyclonal Antibody detects endogenous levels of Integrin beta 5 protein.

Integrin alpha-V/beta-5 (ITGAV:ITGB5) is a receptor for fibronectin. It recognizes the sequence R-G-D in its ligand.; (Microbial infection) Integrin ITGAV:ITGB5 acts as a receptor for adenovirus type C. {ECO:0000269; PubMed:20615244}.
Specifications

Specifications

ITGB5
Polyclonal
Unconjugated
ITGB5
Liquid
ITGB5
Synthesized peptide derived from the C-terminal region of human Integrin Integrin B5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P18084, O70309
INTEGRIN BETA-5, ITGB5,
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16419, 257645, 3693
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit