Learn More
Integrin beta 7 Monoclonal Antibody (FIB504), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc I1141100MG


Description
Specificity: Clone FIB504 recognizes an epitope on human/mouse integrin Beta7. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Mass SpectrometrySpecifications
Integrin beta 7 | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P26010, P26011 | |
Rat | |
Protein A/G | |
RUO | |
16421, 3695 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Mass Spectrometry | |
FIB504 | |
Functional Grade | |
ITGB7 | |
Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen | |
TK1 Cells | |
100 mg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.