Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 7 Polyclonal Antibody
Integrin beta 7 Polyclonal Antibody
Supplier: Thermo Scientific PA547649
Description
Integrin beta 7 Polyclonal Antibody for Western Blot, Flow

Specifications
Integrin beta 7 | |
Polyclonal | |
Unconjugated | |
ITGB7 | |
Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
3695 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P26010 | |
ITGB7 | |
CHO-derived recombinant human Integrin beta 7 long isoform Glu20-His723. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction