Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 7, Rabbit anti-Human,Mouse, Clone: FIB504, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen Integrin beta 7
Clone FIB504
Applications Flow Cytometry, Immunofluorescence, Blocking Assay
Host Species Rabbit
Target Species Human, Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015717
View Documents
Novus Biologicals
NBP2810860.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015718
View Documents
Novus Biologicals
NBP2810860.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human, Mouse Samples. It is validated for Flow Cytometry, Immunofluorescence, Blocking Assay.
Specifications

Specifications

Integrin beta 7
Flow Cytometry, Immunofluorescence, Blocking Assay
Human, Mouse
Primary
FIB504
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit