Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 8 Antibody (2723C), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB47752M
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Integrin beta 8 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
2723C | |
Flow Cytometry, Immunocytochemistry | |
integrin beta-8, integrin, beta 8 | |
Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 8, Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 8 (Glu43-Arg684), Accession # NP_002201.1 | |
0.1 mL | |
Signal Transduction | |
3696 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction