Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 8 Mouse anti-Human, Alexa Fluor 350, Clone: 416922, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB4775U100UG

Catalog No. FAB4775U100


Add to cart
This item is not returnable. View return policy

Description

Description

Integrin beta 8 Monoclonal specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Integrin beta 8
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P26012
ITGB8
Mouse myeloma cell line NS0-derived recombinant human Integrin beta 8 Glu43-Arg684 (predicted) Accession # P26012
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Integrin beta 8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Integrins beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 7, or recombinant mouse Integrin beta 8 is observed.
Flow Cytometry
416922
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
integrin beta-8, integrin, beta 8, ITGB8
Mouse
IgG2b
RUO
Primary
3696
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)