Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 8 Mouse anti-Human, Alexa Fluor 488, Clone: 416922, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4775G100UG
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin beta 8 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P26012 | |
ITGB8 | |
Mouse myeloma cell line NS0-derived recombinant human Integrin beta 8 Glu43-Arg684 (predicted) Accession # P26012 | |
100 μg | |
Primary | |
Detects human Integrin beta 8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Integrins beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 7, or recombinant mouse Integrin beta 8 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
416922 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
integrin beta-8, integrin, beta 8, ITGB8 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3696 | |
Human | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)