Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin pan-αV Rabbit anti-Human, Unconjugated, Clone: EM01309, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Integrin pan-αV
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB118925MI
View Documents
MilliporeSigma
ZRB118925UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11894XMI
View Documents
MilliporeSigma
ZRB11894X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Integrin pan-αV Monoclonal antibody specifically detects Integrin pan-αV in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

Integrin pan-αV
Unconjugated
Rabbit
Vitronectin receptor;Vitronectin receptor subunit alpha;CD51
Human recombinant transmembrane-truncated (DTM) extracellular domains of integrins avb3, avb5, avb6, and avb8
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Purified
Human
ITGAV, MSK8, VNRA, VTNR
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit