Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αvβ6 Mouse anti-Human, R718, Clone: 10D5, BD Horizon™
Mouse Monoclonal Antibody
$188.00 - $458.00
Specifications
Antigen | Integrin αvβ6 |
---|---|
Clone | 10D5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Integrin αvβ6 Monoclonal antibody specifically detects Integrin αvβ6 in Human samples. It is validated for Flow cytometrySpecifications
Integrin αvβ6 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
129/C57BL/6 β6 -/- Mice were mmunized with Human αvβ6 or Mouse Keratinocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
10D5 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
αVβ6; avb6; CD51/β6; ITGAV/ITGB6; Integrin alpha v beta 6 | |
IgG2a | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title