Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αVβ8 Rabbit anti-Human, Unconjugated, Clone: EM13309, MilliporeSigma™

Rabbit Monoclonal Antibody

Manufacturer:  MilliporeSigma ZRB11924X25UL

 View more versions of this product

Catalog No. ZRB11924XMI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Integrin αVβ8 Monoclonal antibody specifically detects Integrin αVβ8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

Integrin αVβ8
EM13309
Unconjugated
Fibronectin Receptor
Rabbit
IgG
Protein A Purified
Signaling
Primary
4X
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
ITGAV:ITGB8
Recombinant fragment corresponding to amino acids 31-987 of integrin alphaV and 43-684 of integrin beta8.
Target Molecular Weight 116.04 and 85.63 kDa calculated (for Integrin alpha V and beta8, respectively).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit