Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αVβ8 Rabbit anti-Human, Unconjugated, Clone: EM13309, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Integrin αVβ8
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB119225MI
View Documents
MilliporeSigma
ZRB119225UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11924XMI
View Documents
MilliporeSigma
ZRB11924X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Integrin αVβ8 Monoclonal antibody specifically detects Integrin αVβ8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

Integrin αVβ8
Unconjugated
Rabbit
Fibronectin Receptor
Recombinant fragment corresponding to amino acids 31-987 of integrin alphaV and 43-684 of integrin beta8.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Purified
Human
ITGAV:ITGB8
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit