Learn More
Integrinalpha2b/CD41 Rat anti-Mouse, Janelia Fluor 549, Clone: MWReg30, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143415JF549
Catalog No. NBP43415549
Add to cart
Description
Description
Integrinalpha2b/CD41 Monoclonal antibody specifically detects Integrinalpha2b/CD41 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Integrinalpha2b/CD41
MWReg30
Flow Cytometry
Purified
ITGA2B
The immunogen for this antibody was CD41.
Protein A or G purified
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary
3674
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Rat
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Mouse