Learn More
IntegrinalphaD/CD11d Mouse anti-Canine, Janelia Fluor 549, Clone: CA11.8H2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10065292JF549
Catalog No. NB065292549
Add to cart
Description
Description
IntegrinalphaD/CD11d Monoclonal antibody specifically detects IntegrinalphaD/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IntegrinalphaD/CD11d
CA11.8H2
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ITGAD
Affinity purified CD11 antigen from canine spleen
Protein A purified
Signal Transduction
Primary
3681
Canine
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp.