Learn More
IntegrinalphaE/CD103 Mouse anti-Human, Janelia Fluor 646, Clone: LF61, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10065272JF646
Catalog No. NB065272646
Add to cart
Description
Description
IntegrinalphaE/CD103 Monoclonal antibody specifically detects IntegrinalphaE/CD103 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IntegrinalphaE/CD103
LF61
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ITGAE
Hairy cell leukemia cells
Protein G purified
Innate Immunity
Primary
3682
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
NB100-65272 recognizes the human CD103 cell surface antigen, a glycoprotein expressed by approximately 1% of peripheral blood lymphocytes, activated T lymphocytes and by hairy cell leukaemia cells. The antigen is also expressed by intraepithelial lymphocytes. It has recently been shown to be identical to the alpha E integrin.