Learn More
IntegrinalphaL/CD11a Rat anti-Mouse, Janelia Fluor 549, Clone: M17/4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141264JF549
Catalog No. NBP14264549
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IntegrinalphaL/CD11aMonoclonalantibody specifically detects IntegrinalphaL/CD11a in Mouse samples.
Specifications
Specifications
IntegrinalphaL/CD11a
M17/4
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ITGAL
C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Protein A purified
Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primary
3683
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Rat
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180 kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.