Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IntegrinalphaV+beta6 Rat anti-Human, Janelia Fluor 549, Clone: 7k, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250448JF549
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IntegrinalphaV+beta6 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
3T3b6, mouse fibroblast expressing human beta6 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
7k | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Rat | |
Protein G purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
Does not bind mouse. | |
Store at 4°C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title