Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrinbeta2/CD18 Rabbit anti-Human, Clone: JM71-31, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266908
Description
Integrinbeta2/CD18 Monoclonal antibody specifically detects Integrinbeta2/CD18 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Integrinbeta2/CD18 | |
JM71-31 | |
Unconjugated | |
Purified | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
ITGB2 | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
Human |
Western Blot,Immunoprecipitation | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunoprecipitation 1:10 - 1:50 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
3689 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title