Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intelectin-1/Omentin Mouse anti-Human, Alexa Fluor™ 532, Clone: 6C11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242652AF532
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Western Blot, ELISA.Specifications
Intelectin-1/Omentin | |
6C11 | |
Western Blot, ELISA | |
Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin | |
Mouse | |
IgG1 Kappa | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Western Blot, ELISA | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ITLN1 | |
Recombinant human ITLN1 (17-313aa) purified from E. coli | |
0.1 mL | |
Primary | |
55600 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title