Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intelectin-1/Omentin Mouse anti-Human, Clone: ITLN1/4061, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Intelectin-1/Omentin
Clone ITLN1/4061
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122828
View Documents
Novus Biologicals
NBP30767720UG
20 μg
Each for $138.37
Add to cart
 
NB122827
View Documents
Novus Biologicals
NBP30767710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Intelectin-1/Omentin Monoclonal antibody specifically detects Intelectin-1/Omentin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Intelectin-1/Omentin
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
55600
IgG2b Kappa
Primary
Store at 4°C.
ITLN1/4061
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin
Recombinant fragment (around aa16-141) of human Intelectin-1/Omentin protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit