Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
The BUCHI Interface I-300 controls all distillation parameters of a Rotavapor R-300 system including the optional V-300 Vacuum Pump and F-3XX series chillers.
Supplier: BUCHI 11I300S20
Description
- Easy plug and play integration into a Rotavapor™ R-300 system at any time
- Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
- Remote monitoring on smart phones including push notifications and live view
- Modes: manual, timer, continuous
- Flexibility of the interface allows easy installation and convenient operation from multiple positions:
- Rotavapor™ handle
- Vacuum pump
- Fume hood or lab bench wall
- Stand-alone operation to locally control a central vacuum system
- Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
- Unattended operation (optional):
- Automatic distillation with CloudDest sensor
- Foam sensor for foaming samples
Specifications
Vacuum Controller/Interface | |
R-300, V-300, V-600 | |
Kit for V-300/600, Woulff Bottle | |
Standard |