Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha K Antibody (OTI2A10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271007AF647
Description
IFN-alpha K Monoclonal specifically detects IFN-alpha K in Human samples. It is validated for Western Blot.Specifications
IFN-alpha K | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IFNA6 | |
Full length recombinant protein of human IFN-alpha K (NP_066282) produced in E.coli. | |
0.1 mL | |
3443.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2A10 | |
Western Blot | |
IFNA6, IFN-alpha 6, IFNalpha K, IFN-alpha K, IFN-kappa, interferon kappa, interferon, kappa, RP11-27J8.1, UNQ6124/PRO20084 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction