Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon-α (II)/IFN-α (II) Mouse Monoclonal Antibody, Epredia™

Epredia™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia MS574PABX

 View more versions of this product

Catalog No. MS574PABX


Add to Cart

Description

Description

The IFN-α family consists of 24 or more genes or pseudo-genes. IFN-αII) is one of the two distinct families (I and II) of human IFN-α. The α interferons are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.

Host Species: Mouse

Clone: N27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 43-53

Immunogen: Purified recombinant IFN-α 2c

Molecular Weight: 16-27kDa

Positive Control: Stimulated human peripheral blood mononuclear cells

Recommended for:

  • Blocking study

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Interferon-alpha (II)/IFN-alpha (II)
N27
Unconjugated
Purified recombinant IFN-alpha 2c
RUO
Primary
Concentrated
Monoclonal
1mg/mL
Mouse
200 μL
Cytokines
Human
IgG2a κ
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit