Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferonalpha Mouse anti-Human, Janelia Fluor 549, Clone: 144BS, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP229374JF549

Catalog No. NBP29374549


Add to cart

Description

Description

Interferonalpha Monoclonal antibody specifically detects Interferonalpha in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Interferonalpha
144BS
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Affinity Purified
IFNA1
This antibody was raised against Interferon alpha.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
This antibody binds but does not neutralize the activity of human IFN-alpha subtypes - B, -C, -D and -F, as well as several IFN-alpha B/D, D/B hybrids.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
IFN alpha
Mouse
IgG1 Kappa
0.1mL
Primary
3439
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit