Learn More
Thermo Scientific Chemicals Interleukin 3, mouse
$123.90
Specifications
Molecular Weight (g/mol) | 15.6 kd |
---|---|
Packaging | Plastic bottle |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ67104AME
|
Thermo Scientific Chemicals
J67104AME |
2μg |
Each for $123.90
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ67104EXE
|
Thermo Scientific Chemicals
J67104EXE |
10μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
15.6 kd | |
Plastic bottle | |
Lyophilized |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.