Learn More
Motic Swift Line Intermediate Compound Microscopes Products for Science Education
Motic Swift Line Intermediate Compound Microscopes set the standard for middle and high school instruction and include all of the most commonly specified microscope features.
$245.00 - $475.00
Specifications
For Use With (Application) Intermediate Science Education
Magnification Power 40X, 100X, 400X
Objective Types Achromatic
View More Specs
Products 4
Catalog Number Mfr. No. Design Type Head Style Lighting Type Price Quantity & Availability  
Catalog Number Mfr. No. Design Type Head Style Lighting Type Price Quantity & Availability  
S81366 Products for Science Education
View Documents
Motic Swift Line
127CLED
Stage: Clips Monocular LED, Corded
Each for $245.00
Add to cart
S81369 Products for Science Education
View Documents
Motic Swift Line
128CLED
Stage: Clips Dual View LED, Corded
Each for $375.00
Add to cart
S81370 Products for Science Education
View Documents
Motic Swift Line
129CLED3
Stage: Mechanical Monocular LED, Corded
Each for $475.00
Add to cart
S81374 Products for Science Education
View Documents
Motic Swift Line
127
Stage: Clips Monocular Tungsten
Each for $250.00
Add to cart
Description
Description
  • Widefield 10X eyepiece with revolving pointer
  • Viewing head rotates 360° for easy repositioning of eyepiece and sharing by more than one student
  • Separate low-position coarse and fine focusing
  • Choice of monocular, binocular, or dual view heads
  • Available with stage clips or mechanical stage (129-CLED-3)
Specifications
Specifications
Intermediate Science Education
40X, 100X, 400X
Achromatic