Learn More
Science First Intermediate Force Table
Classic way to study force and vectors.
$21.50
Specifications
Includes | Force table, four low-friction pulleys (S409673A), four slotted weight hangers, tripod base, instructions with theory and sample problem |
---|
Includes: Force table, four low-friction pulleys (S409673A), four slotted weight hangers, tripod base, instructions with theory and sample problem
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S65116
![]() |
Science First
6110095 |
Force table paper |
Pack of 50 for $21.50
|
|
Description
- Lightweight, accurate and simple
- Permanently printed decal with large, easy-to-read numerals is graduated in two 360° scales to prevent parallax error
- Masonite, 12 in. dia. (40cm) disc balances on sturdy cast-iron tripod base which can be fine tuned with leveling feet
- Weight harness snaps into place around a central ring
Order Info
Double 360° scale decal on 12 in. dia. (40cm) replacement paper displays calculations or notes during lab presentation. Discard paper or display as part of the experiment. Sold in packs of 50 sheets.
Specifications
Force table, four low-friction pulleys (S409673A), four slotted weight hangers, tripod base, instructions with theory and sample problem |