Learn More
International Enviroguard™ MicroGuard CE™ Lab Coats
Utilizes state-of-the-art lamination technology to combine a soft nonwoven fabric with a microporous film
$370.50 - $431.50
Specifications
Certifications/Compliance CE
Collar Type Bound neck
Closure Type Snaps
Color White
Cuff Style Tunneled elastic
View More Specs
Products 12
Catalog Number Mfr. No. Length (English) Size Packaging Type Size (English) Chest Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Packaging Type Size (English) Chest Price Quantity & Availability  
19-472-531 View Documents International Enviroguard™
CE8026CIM
38.5 in. Medium Clean Processed; Individually Wrapped 38 in. Case of 50 for $370.50
Add to cart
19-472-532 View Documents International Enviroguard™
CE8026CIL
40 in. Large Clean Processed; Individually Wrapped 40 in. Case of 50 for $370.50
Add to cart
19-472-533 View Documents International Enviroguard™
CE8026CIXL
41 in. X-Large Clean Processed; Individually Wrapped 42 in. Case of 50 for $370.50
Add to cart
19-472-534 View Documents International Enviroguard™
CE8026CI2XL
42 in. 2X-Large Clean Processed; Individually Wrapped 44 in. Case of 50 for $388.00
Add to cart
19-472-536 View Documents International Enviroguard™
CE8026CI4XL
44.5 in. 4X-Large Clean Processed; Individually Wrapped 46 in. Case of 50 for $431.50
Add to cart
19-472-535 View Documents International Enviroguard™
CE8026CI3XL
43.25 in. 3X-Large Clean Processed; Individually Wrapped 45 in. Case of 50 for $412.50
Add to cart
19-472-528 View Documents International Enviroguard™
CE8026BP2XL
42 in. 2X-Large Bulk packaged 44 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-472-526 View Documents International Enviroguard™
CE8026BPL
40 in. Large Bulk packaged 40 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-472-527 View Documents International Enviroguard™
CE8026BPXL
41 in. X-Large Bulk packaged 42 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-472-525 View Documents International Enviroguard™
CE8026BPM
38.5 in. Medium Bulk packaged 38 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-472-529 View Documents International Enviroguard™
CE8026BP3XL
43.25 in. 3X-Large Bulk packaged 45 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-472-530 View Documents International Enviroguard™
CE8026BP4XL
44.5 in. 4X-Large Bulk packaged 46 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Bound seams to eliminate raw edges and particulate generation
  • Tunnelized elastic is encapsulated to provide secure closures
  • Low linting
  • High Moisture Vapor Transmission Rate (MVTR)
  • Liquid Barrier
  • Economical and recyclable
Appropriate for cleanrooms classified as ISO9 through ISO5
Order Info
Available in bulk packages or individually wrapped clean process packages
Specifications
Specifications
CE
Snaps
Tunneled elastic
Serged
Bound neck
White
Non-woven fabric