Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ PVC Cleanroom Chemical Aprons with Long Sleeves
Click to view available options
Size:
Medium
X-Large
2X-Large
Description
- Heavy-duty, 6 mil thick chemical-resistant aprons are made of PVC
- Designed to protect against acids, alcohol, oils, grease, fats, and a variety of chemicals
- Liquid-resistant, abrasion-resistant, and puncture-resistant to withstand tough work environments
- Protect against liquid splash, sprays, and non-hazardous dry particulates
- Loop over the head and nylon ties on the back
- Edges are ultrasonically welded to reduce linting and release of unwanted particulates
- Offer full protection of the front of the body from the chest to the knees or below
- Long sleeves with tunnelized elastic cuffs to protect the arms from splashes and sprays and to also increase particle containment
- Cleanroom-ready aprons are clean-processed and individually bagged
- Applications include semi-conductor, cleanroom, pharmaceutical, chemical processing, petrochemicals, sanitation work
- Latex-free elastic
Specifications
Type | Controlled Environments Apron |
Size | 2X-Large |
Material | PVC |
Color | Blue |
Disposable | Single-use |
Closure Type | Waist Ties |
Sterility | Non-sterile |
Resistance | Liquid, Abrasion, Puncture |
Thickness | 6 mil |
For Use With (Application) | Semi-conductor, Cleanroom, Pharmaceutical, Chemical Processing, Petrochemicals, Sanitation Work |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction