Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Intertek™ MIC Test Kit, Petroleum Microbiology Media: Acid Producing Bacteria Media
Microbiology growth media products
Manufacturer:  INTERTEK WESTPORT TECHNOLOGY CENTER MK108A
 View more versions of this product
Catalog No. 08-629-016A
Description
Description
A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Acid Producing Bacteria (APB) contribute to MIC by decreasing the pH of the environment which promotes the dissolution of metal.
  • APB: for quantification of acid producing bacteria and general heterotrophic anaerobic bacteria
  • The change in pH (production of acid) in APB-Q media is indicated by a change in color from red to yellow
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
United States
1% salinity
APB-Q, 1% salinity
Red
Tube
12 Tubes
Cultivation of Acid producing bacteria, MIC testing