Learn More
Intertek™ MIC Test Kit, Petroleum Microbiology Media: HAB
Quantification of heterotrophic (general) aerobic bacteria
Manufacturer:  INTERTEK WESTPORT TECHNOLOGY CENTER MK122C
 View more versions of this product
Catalog No. 08-629-007C
Add to cart
Description
Description
For use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
Vial
12 Vials
Cultivation of aerobic bacteria, MIC testing
United States
6% salinity
HAB, 6% salinity