Learn More
Intertek™ MIC Test Kit, Petroleum Microbiology Media: SBN
Standard Bacteriological Nutrient medium, for anaerobic and facultative anaerobic heterotrophic bacteria
Manufacturer:  INTERTEK WESTPORT TECHNOLOGY CENTER MK119C
 View more versions of this product
Catalog No. 08-629-012C
Add to cart
Description
Description
For use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
Vial
288 Vials
Cultivation of anaerobic and facultative anaerobic heterotrophic bacteria, MIC testing
United States
salinity (6%)
SBN, 6% salinity