Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intertek™ MIC Test Kit, Petroleum Microbiology Media: SRB/APB Combo Kits

Microbiology growth media products

Manufacturer:  INTERTEK WESTPORT TECHNOLOGY CENTER MK109A

 View more versions of this product

Catalog No. 08-629-002A


Add to cart

Description

Description

A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC).

Sulfate Reducing Bacteria (SRB) contribute to MIC by producing hydrogen sulfide, which is a corrosive and toxic gas that has the odor of rotten eggs. SRB can also sour oil and gas reservoirs, create precipitates that plug openings and discolor water to black. Acid Producing Bacteria (APB) contribute to MIC by decreasing the pH of the environment which promotes the dissolution of metal.

  • Five vials SRB media for the enumeration of sulfate-reducing bacteria
  • Five vials APB (red) media for the enumeration of acid producing bacteria
  • One bottle sterile diluent (D) for making suspensions of biofilm, sludge, scale, etc.
  • One vial 2% formalin (P) (red top) for preserving water/solids for microscopic examination
  • Five syringes + 5 needles for performing inoculations and serial dilution steps
  • One swab + 1 wooden scraper for collecting biofilm, deposits, and sludge
  • One alcohol pad for sterilizing tops of media vials

Compliance

Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.

Specifications

Specifications

NACE, TM0194-94
Vial
1 Kit
MIC testing
United States
1% salinity
SRB, APB Combi Kit, 1% salinity
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit