Intertek™ MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media

Microbiology growth media products

Manufacturer: INTERTEK WESTPORT TECHNOLOGY CENTER MK102B

 View more versions of this product

Catalog No. 08-629-015B

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 


    Specifications

    Certifications/Compliance NACE, TM0194-94
    Color Clear
    For Use With (Application) Cultivation of Sulfate reducing bacteria, MIC testing
    Manufacturing Origin United States
    Media Type SRB-Q, 2.5% salinity
    Packaging Tube
    Product Type SRB-Q, 2.5% salinity
    Quantity 12 Tubes

    A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.

    Sulfate Reducing Bacteria (SRB) contribute to MIC by producing hydrogen sulfide, which is a corrosive and toxic gas that has the odor of rotten eggs. SRB can also sour oil and gas reservoirs, create precipitates that plug openings and discolor water to black.

    Sulfate Reducing Bacteria Media

    • SRB: for quantification of sulfate reducing bacteria (Modified Postgates B Medium)
    • SRB-Q: quick format single tube test for SRB
    • SRB2: specially formulated clear SRB media
    • SRB-Q media provides a convenient way to estimate the concentration of SRB in water samples by changing the clarity of the color of the media from white to black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit