Intertek™ MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media

Microbiology growth media products

Manufacturer: INTERTEK WESTPORT TECHNOLOGY CENTER MK103A

 View more versions of this product

Catalog No. 08-629-019A

  Check Availability
Add to Cart

    Specifications

    Manufacturing Origin United States
    Packaging Vial
    Product Type SRB2, 1% salinity
    Quantity 12 Vial
    Color Clear
    For Use With (Application) Cultivation of Sulfate reducing bacteria, MIC testing
    Media Type SRB2, 1% salinity
    Certifications/Compliance NACE, TM0194-94

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit