Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA126880
Description
PA1-26880 detects calf intestinal Alkaline Phosphatase from bovine samples.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
Specifications
| Intestinal Alkaline Phosphatase | |
| Polyclonal | |
| HRP | |
| ALPI | |
| 2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097 | |
| Sheep | |
| IgG fraction | |
| RUO | |
| 280993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot, ELISA, Western Blot | |
| 10 mg/mL | |
| PBS with 1% BSA, 0.15M NaCl and 0.01% gentamicin sulfate; pH 7.2 | |
| P19111 | |
| ALPI | |
| Alkaline Phosphatase collected from calf Intestine. | |
| 1 mg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction