Learn More
BioVision IncSupplier Diversity Partner EZCell™ Intracellular Nitric Oxide Synthase (NOS) Detection Kit: Cell Based simple, one step, non-radiometric method for detection of intracellular NOS / Nitric Oxide Synthase. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K207100
EZCell™ Intracellular Nitric Oxide Synthase (NOS) Detection Kit: Nitric oxide synthases (EC 1.14.13.39) (NOSs) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). nNOS accounts for the production of NO in central nervous system, where NO participates in cell communication and information storage. eNOS produces NO in blood vessels and is involved with the regulation of vascular function. BioVision's Intracellular Nitric Oxide Synthase Detection kit uses a dye that reacts with intracellular NO produced by NOS to produce fluorescence (Ex/Em = 485/530 nm), which is proportional to the concentration of intracellular NOS.This kit provides a simple, non-radiometric method for detection of intracellular NOS in the cells.
Catalog No. 50-111-9619