Learn More
Cell Biolabs IncSupplier Diversity Partner INTRACELLULAR ROS KT 96WL  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA342T
OxiSelect Intracellular ROS Assay Kit (Trial Size) is a cell-based assay for measuring reactive oxygen species within a cell. The assay employs a cell-permeable fluorogenic probe that detects a variety of ROS. 20 assays/kit.
Catalog No. 50-673-534