Learn More
Thermo Scientific INTRO SAMPLE KIT DFS Item
Manufacturer:  Thermo Scientific 842312051791
Radial Standard Kit; Kit consisting of an EMT torch holder, EMT centre tube holder, radial; EMT torch, 1.5 mm glass centre tube, radial spray chamber adaptor, glass; spray chamber, concentric; glass nebulizer, ball joint clip, a pack of 6 aqueous sample tubes and; a pack of 6 aqueous drain tubes.; The kit is presented in a purpose designed case and is intended for; standard aqueous analysis applications and is suitable only with radial; ICP systems.
Catalog No. NC0053976