Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Introduction to Chromatography Lab Products for Science Education

Separate plant pigments and dye samples to learn about chromatography in clinical, forensic and analytical laboratories.

Specifications

Class Size 40 students
For Use With (Application) Separates Plant Pigments and Dye Samples, Employed to Identify Organic and Inorganic Compounds
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S67396 Products for Science Education
View Documents
Neo/Sci Corporation
201683
Lab Investigation Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • See the use of chromatography in identifying organic and inorganic compounds
  • Learn how solvent and substrate variables affect chromatography
  • Calculate the Rf values of unknown dye samples

Order Info

Class size: 40 students.

Specifications

Specifications

40 students
Separates Plant Pigments and Dye Samples, Employed to Identify Organic and Inorganic Compounds
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit