Learn More
Lab-Aids™ Introduction to Conductivity Products for Science Education
Explores electrical conductivity, develop principles of conductors and insulators and electrolytes/non-electrolytes using acids, bases, salts, alcohols, and hydrocarbons, test a variety of common materials for their conductivity characteristics and differentiate between strong, weak, and non-conductors of electricity.
Manufacturer:  Lab-Aids™ 309
Catalog No. S20663
Add to cart
Includes: One teachers guide with MSDS; 30 student worksheets and guides; 15 Chemplates with measuring and mixing spatulas; 1 string, cut, pkg; 1 paper clips, metal, pkg; 1 pics, plastic, pkg; 1 rubber bands, pkg; 1 insulation copper wire, strips, pkg; 2 bottles hydrochloric acid, 6M, 0.25 oz.; 2 bottles denatured alcohol, 0.25 oz; 2 bottles glacial acetic acid, 0.25 oz.; 3 bottles deionized water, 2 oz.; 1 vial calcium hydroxide; 1 vial sodium chloride crystals; 1 vial sucrose crystals, solid
Description
Description
All materials can be classified as a conductor, insulator or semi-conductor. Materials are classified according to their resistance to the flow of current. A conductor is a substance that is capable of carrying an electric charge from one point to another. An insulator blocks the flow of current. Semi-conductors are neither. Their conductivity lies in between the two.
In Part 1 of this kit, students test several common objects to determine if they are conductors or insulators.
In Part 2, The students test electrical conductivity in aqueous solutions. They test an acid, base, salt, alcohol and a hydrocarbon.
This kit can be used as an introduction for students in physical science, general science and introductory chemistry courses. Conductivity indicators are required for these activities, but are not included with this kit. This kit is packaged complete for 30 students.
Specifications
Specifications
Kit: Introduction to Conductivity
Education
For 30 students/15 groups
One teachers guide with MSDS; 30 student worksheets and guides; 15 Chemplates with measuring and mixing spatulas; 1 string, cut, pkg; 1 paper clips, metal, pkg; 1 pics, plastic, pkg; 1 rubber bands, pkg; 1 insulation copper wire, strips, pkg; 2 bottles hydrochloric acid, 6M, 0.25 oz.; 2 bottles denatured alcohol, 0.25 oz; 2 bottles glacial acetic acid, 0.25 oz.; 3 bottles deionized water, 2 oz.; 1 vial calcium hydroxide; 1 vial sodium chloride crystals; 1 vial sucrose crystals, solid