Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTS1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550608

Catalog No. PA550608


Add to cart

Description

Description

The antibody detects endogenous levels of total INTS1 protein.

INTS1 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2. RNA polymerase II (Pol II) is an enzyme that is composed of twelve subunits and is responsible for the transcription of protein-coding genes. Transcription initiation requires Pol II-mediated recruitment of transcription machinery to a target promoter, thereby allowing transcription to begin.
Specifications

Specifications

INTS1
Polyclonal
Unconjugated
INTS1
Liquid
INTS1
Synthetic peptide corresponding to residues near the N terminal of human integrator complex subunit 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8N201, Q6P4S8
KIAA1440
Rabbit
IgG
100 μL
-20°C
Primary
26173, 68510
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit