Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS10 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305046100UL
Description
INTS10 Polyclonal specifically detects INTS10 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
INTS10 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
55174 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
C8orf35, INT10, integrator complex subunit 10 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 451-710 of human INTS10 (NP_060612.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction