Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen INTS6
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092709
View Documents
Novus Biologicals
NBP304624100UL
100 μg Each for $320.71
Add to cart
 
NB092710
View Documents
Novus Biologicals
NBP30462420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

INTS6 Polyclonal antibody specifically detects INTS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

INTS6
Unconjugated
Rabbit
Cancer, Growth and Development, Neuronal Cell Markers, Tumor Suppressors
PBS with 50% glycerol, pH7.3.
26512
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse, Rat
DBI1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26, DICE1DBI-1DDX26ADDX26DEAD box protein, DKFZp434B105, HDB, INT6, integrator complex subunit 6, Notchl2, Protein DDX26, Protein deleted in cancer 1, RNA helicase HDB
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human INTS6 (NP_036273.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit