Learn More
INTS8 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188021AP
Catalog No. 50-173-2230
Add to cart
Description
Description
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
INTS8
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q75QN2
C8orf52, INT8, integrator complex subunit 8, INTS8, Protein kaonashi 1
Rabbit
IgG
150 μL
-20°C
Primary
55656
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
INTS8
Liquid
INTS8
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human