Alfa Aesar™ Invar™, Iron Nickel powder, -325 mesh, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4460022

Quantity 100g

 View more versions of this product

Catalog No. AA4460022

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01741928
    UN Number UN3089
    Chemical Name or Material Invar™ Iron Nickel powder

    Specifications

    Quantity 100g
    Health Hazard 2 GHS H Statement
    H228-H372-H351-H319-H317-H335
    Flammable solid.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD01741928
    Odor Odorless
    TSCA TSCA
    UN Number UN3089
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Mesh Size −325 Mesh
    Physical Form Powder
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Ni 65:35 wt%
    Color Gray
    Chemical Name or Material Invar™ Iron Nickel powder
    Assay 99.9%, (metals basis)

    GHS H Statement
    H228-H372-H351-H319-H317-H335
    Flammable solid.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit