Learn More
Investigating Animal Behavior
Study the response of organisms to environmental conditions.
Specifications
For Use With (Application) | Students use specially molded choice chambers to study the response of organisms to environmental conditions |
---|---|
Includes | Eight choice chambers and prepared solutions to test various environmental conditions, additional choice chambers available |
Includes: Eight choice chambers and prepared solutions to test various environmental conditions; additional choice chambers available
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S66652
![]() |
Neo/Sci Corporation
202933 |
Investigate Animal Behavior Lab |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
In this lab investigation, students use specially molded choice chambers to study the response of organisms to environmental conditions.
- Determine the environmental preference of locally available animals, such as sow bugs, ants, earthworms and other small invertebrates
- Test students' predictions regarding habitat preference, as well as the innate and learned behaviors of animals
Specifications
Students use specially molded choice chambers to study the response of organisms to environmental conditions | |
Eight choice chambers and prepared solutions to test various environmental conditions, additional choice chambers available |