Learn More
Lab-Aids™ Investigating the Design & Output of Wet Cell Batteries
SCI_ED
Design and conduct a scientific investigation, use appropriate tools and techniques to gather, analyze, and interpret data and use evidence to develop descriptions, explanations, predictions, and models.
Supplier:  Lab-Aids™ 205S
Catalog No. S20954
Add to cart
Includes: Teacher's guide with MSDS; 28 student worksheets and guides; 30 table salt; 12 strips each: copper, iron, magnesium, zinc; 6 electric motors; 6 Wet Cell Chambers (SEPUP); 6 black wire leads with alligator clips; 6 3% Hydrogen Peroxide solution, 30mL dropper bottles; 6 graduated cups, 30mL; 6 plastic cups, 270mL; 6 sandpaper, small pieces; 6 red wire leads with alligator clips; 6 plastic spoons; Transparency: Sample Procedure and Data Table for Part B; Transparency: Metals Activity Chart
Description
Description
This kit introduces students to the concept of energy conversion through the study of a simple electrochemical cell connected to an electric motor. Electrochemical cells are the basis for batteries and contain metals and other chemicals with stored chemical potential energy. Students design and conduct an experiment that uses the unique Lab-Aids™ Wet Cell Chamber to determine which combination of different metals works best to convert the potential chemical energy into electric energy. Their results allow them to rank the metals tested in order of reactivity. They also investigate how the distance between the metals, the direction of current flow, and the amount of metal surface area impacts the electrical output of the wet cell. Accommodates 6 groups of 4 students. Includes Teacher guide, student sheets, assessment questions and SDS for all chemicals.
Specifications
Specifications
Teacher's guide with MSDS; 28 student worksheets and guides; 30 table salt; 12 strips each: copper, iron, magnesium, zinc; 6 electric motors; 6 Wet Cell Chambers (SEPUP); 6 black wire leads with alligator clips; 6 3% Hydrogen Peroxide solution, 30mL dropper bottles; 6 graduated cups, 30mL; 6 plastic cups, 270mL; 6 sandpaper, small pieces; 6 red wire leads with alligator clips; 6 plastic spoons; Transparency: Sample Procedure and Data Table for Part B; Transparency: Metals Activity Chart
Kit: Wet Cell Batteries
24 Students (6 Groups)