Learn More
Investigating Energy Transfer
This lesson consists of two experiments: “Exploring Radiation and Absorption” and “Exploring Conduction.”
Supplier: Science First 6231055
Requires : Three thermometers, lamp with reflector, ring stand, paper towels, tape, hot water, cold water
Description
The instructor's manual lists Benchmarks for Science Literacy and National Science Education Standards.
Exploring Radiation and Absorption
- Observe how surface color affects rate of energy absorption by an object
- Observe how surface color affects rate of energy radiation from an object
Exploring Conduction
- Measure temperature change in two containers connected by a metal bar
- Observe how conduction affects the temperature of objects in a system
Specifications
Investigating Energy Transfer | |
The instructor's manual lists |